Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbs5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108583.2 | NP_001102053.1 | 8 - 9 | 713 | 133 | |
GenBank mRNA | BC167024.1 | - | 8 - 9 | 713 | 7123978 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |