Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serinc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110811.1 | NP_001104281.1 | 1 - 2 | 423 | 71 | |
XM_006234853.2 | XP_006234915.1 | 1 - 2 | 505 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |