Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddrgk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106512.1 | NP_001099982.1 | 3 - 4 | 843 | 86 | |
GenBank mRNA | FQ214225.1 | - | 3 - 4 | 428 | 86 | |
FQ231424.1 | - | 3 - 4 | 467 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |