Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldlr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175762.2 | NP_786938.1 | 14 - 15 | 2290 | 60 | |
GenBank mRNA | X13722.1 | - | 14 - 15 | 2290 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |