Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1c12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170342.1 | NP_001163813.1 | 4 - 5 | 493 | 64 | |
XM_006254190.2 | XP_006254252.1 | 6 - 7 | 1343 | 64 | ||
XM_006254191.1 | XP_006254253.1 | 7 - 8 | 849 | 64 | ||
XM_008771873.1 | XP_008770095.1 | 6 - 7 | 733 | 64 | ||
XM_008771874.1 | XP_008770096.1 | 6 - 7 | 659 | 64 | ||
GenBank mRNA | EX487988.1 | - | 4 - 5 | 493 | 64 |
Gene Symbol: Akr1c12l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135744.1 | NP_001129216.1 | 4 - 5 | 481 | 64 | |
GenBank mRNA | BC129122.1 | - | 4 - 5 | 481 | 7127211 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |