Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044247.1 | NP_001037712.1 | 1 - 2 | 263 | 67 | |
XM_008771246.1 | XP_008769468.1 | 1 - 2 | 696 | 67 | ||
XM_008771247.1 | XP_008769469.1 | 1 - 2 | 291 | 67 | ||
GenBank mRNA | BC098935.1 | - | 1 - 2 | 263 | 7445963 | 67 |
FQ214804.1 | - | 1 - 2 | 213 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |