Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039336.1 | NP_001034425.1 | 5 - 6 | 609 | 146 | |
XM_006252072.2 | XP_006252134.1 | 5 - 6 | 659 | 146 | ||
XM_006252073.2 | XP_006252135.1 | 5 - 6 | 859 | 146 | ||
XM_008770749.1 | XP_008768971.1 | 5 - 6 | 674 | 146 | ||
XM_008770750.1 | XP_008768972.1 | 5 - 6 | 860 | 146 | ||
GenBank mRNA | AY886539.1 | - | 5 - 6 | 609 | 146 | |
BC098018.1 | - | 5 - 6 | 573 | 146 | ||
BC127440.1 | - | 5 - 6 | 619 | 7456492 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |