Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gzmn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006252044.2 | XP_006252106.2 | 9 - 10 | 1111 | 73 | |
XM_008770696.1 | XP_008768918.1 | 8 - 9 | 1094 | 73 | ||
XM_008770697.1 | XP_008768919.1 | 9 - 10 | 1162 | 73 | ||
XM_008770698.1 | XP_008768920.1 | 9 - 10 | 1137 | 73 | ||
XM_008770699.1 | XP_008768921.1 | 8 - 9 | 1095 | 73 | ||
XM_008770700.1 | XP_008768922.1 | 6 - 7 | 879 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis Protease 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |