Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079689.2 | NP_001073157.1 | 5 - 6 | 901 | 112 | |
NM_001270846.1 | NP_001257775.1 | 6 - 7 | 550 | 112 | ||
NM_001270847.1 | NP_001257776.1 | 5 - 6 | 449 | 112 | ||
NM_001270848.1 | NP_001257777.1 | 5 - 6 | 534 | 112 | ||
XM_008769542.1 | XP_008767764.1 | 4 - 5 | 830 | 112 | ||
GenBank mRNA | DQ286232.1 | - | 4 - 5 | 427 | 112 | |
DQ923317.1 | - | 5 - 6 | 534 | 112 | ||
DQ923318.1 | - | 5 - 6 | 450 | 112 | ||
DQ923319.1 | - | 6 - 7 | 550 | 112 | ||
DQ923320.1 | - | 5 - 6 | 892 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |