Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191929.1 | NP_001178858.1 | 3 - 4 | 439 | 58 | |
XM_006250187.2 | XP_006250249.1 | 5 - 6 | 639 | 58 | ||
XM_006250188.2 | XP_006250250.1 | 5 - 6 | 635 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Protein Phosphatase 3' Most Recommended Assay Rattus norvegicus Probe spans exons |