Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcrla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100682.1 | NP_001094152.1 | 5 - 6 | 1073 | 72 | |
GenBank mRNA | BC105902.1 | - | 5 - 6 | 1044 | 72 | |
BC158820.1 | - | 5 - 6 | 1073 | 7375319 | 72 | |
FQ225304.1 | - | 5 - 6 | 1060 | 72 | ||
FQ227783.1 | - | 5 - 6 | 1062 | 72 | ||
FQ232269.1 | - | 5 - 6 | 1060 | 72 | ||
FQ233227.1 | - | 5 - 6 | 1059 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |