Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdrd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134739.1 | NP_001128211.1 | 9 - 10 | 1762 | 97 | |
NM_001134740.1 | NP_001128212.1 | 9 - 10 | 1762 | 97 | ||
XM_006250037.2 | XP_006250099.1 | 10 - 11 | 2026 | 97 | ||
XM_006250038.2 | XP_006250100.1 | 10 - 11 | 1844 | 97 | ||
XM_006250039.2 | XP_006250101.1 | 10 - 11 | 1795 | 97 | ||
XM_008769615.1 | XP_008767837.1 | 10 - 11 | 1813 | 97 | ||
XM_008769616.1 | XP_008767838.1 | 10 - 11 | 1980 | 97 | ||
GenBank mRNA | BC168218.1 | - | 9 - 10 | 1762 | 7319314 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |