Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Commd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004276.1 | NP_001004276.1 | 5 - 6 | 528 | 99 | |
GenBank mRNA | BC079373.1 | - | 5 - 6 | 528 | 7110105 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |