Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053905.1 | NP_446357.1 | 2 - 3 | 849 | 159 | |
GenBank mRNA | U72353.1 | - | 2 - 3 | 849 | 159 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |