Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ing4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079887.1 | NP_001073356.1 | 3 - 4 | 279 | 79 | |
XM_006237349.2 | XP_006237411.1 | 3 - 4 | 378 | 79 | ||
XM_006237350.2 | XP_006237412.1 | 3 - 4 | 378 | 79 | ||
GenBank mRNA | BC086570.1 | - | 3 - 4 | 279 | 79 | |
BC097942.1 | - | 3 - 4 | 280 | 79 | ||
BC127511.1 | - | 3 - 4 | 279 | 7376723 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |