Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749821.3 | XP_003749869.1 | 2 - 3 | 636 | 108 | |
XM_008763272.1 | XP_008761494.1 | 2 - 3 | 636 | 108 | ||
XM_575654.6 | XP_575654.3 | 2 - 3 | 636 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |