Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ltbr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008315.1 | NP_001008316.1 | 9 - 10 | 1200 | 121 | |
XM_006237351.1 | XP_006237413.1 | 9 - 10 | 1247 | 121 | ||
GenBank mRNA | BC085880.1 | - | 9 - 10 | 1200 | 7191913 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |