Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012218.1 | NP_001012218.1 | 1 - 2 | 302 | 58 | |
XM_006232181.2 | XP_006232243.1 | 1 - 2 | 340 | 58 | ||
XM_006232182.2 | XP_006232244.1 | 1 - 2 | 340 | 58 | ||
GenBank mRNA | BC087100.1 | - | 1 - 2 | 302 | 7192411 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |