Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044696.1 | NP_001038161.1 | 8 - 9 | 1037 | 141 | |
XM_006232235.2 | XP_006232297.1 | 9 - 10 | 1008 | 141 | ||
XM_008760937.1 | XP_008759159.1 | 9 - 10 | 1099 | 141 | ||
XM_008760938.1 | XP_008759160.1 | 8 - 9 | 795 | 141 | ||
XM_008760939.1 | XP_008759161.1 | 8 - 9 | 991 | 141 | ||
GenBank mRNA | BC097432.1 | - | 8 - 9 | 1037 | 7455342 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |