Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044696.1 | NP_001038161.1 | 9 - 10 | 1194 | 67 | |
XM_006232235.2 | XP_006232297.1 | 10 - 11 | 1165 | 67 | ||
XM_008760937.1 | XP_008759159.1 | 10 - 11 | 1256 | 67 | ||
XM_008760938.1 | XP_008759160.1 | 9 - 10 | 952 | 67 | ||
XM_008760939.1 | XP_008759161.1 | 9 - 10 | 1148 | 67 | ||
GenBank mRNA | BC097432.1 | - | 9 - 10 | 1194 | 7455342 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |