Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfhx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191702.1 | NP_001178631.1 | 6 - 7 | 3866 | 97 | |
XM_006232162.2 | XP_006232224.1 | 6 - 7 | 3972 | 97 | ||
XM_006232164.2 | XP_006232226.1 | 6 - 7 | 3837 | 97 | ||
XM_008760867.1 | XP_008759089.1 | 6 - 7 | 4332 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |