Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031526.1 | NP_113714.1 | 1 - 2 | 225 | 56 | |
GenBank mRNA | X14211.1 | - | 1 - 2 | 130 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Inventoried Probe spans exons |