Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcgf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105836.2 | NP_001099306.1 | 7 - 8 | 712 | 96 | |
XM_008767971.1 | XP_008766193.1 | 8 - 9 | 671 | 96 | ||
XM_008767972.1 | XP_008766194.1 | 8 - 9 | 947 | 96 | ||
XM_008767973.1 | XP_008766195.1 | 8 - 9 | 1017 | 96 | ||
GenBank mRNA | BC167059.1 | - | 7 - 8 | 712 | 7445511 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |