Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013854.1 | NP_001013876.2 | 3 - 4 | 773 | 72 | |
NR_073160.1 | 3 - 4 | 780 | 72 | |||
GenBank mRNA | BC079025.1 | - | 3 - 4 | 773 | 7113945 | 72 |
FQ213681.1 | - | 3 - 4 | 572 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons |