Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fndc3c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191722.1 | NP_001178651.1 | 2312 | 99 |
Gene Symbol: LOC103694495 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773355.1 | XP_008771577.1 | 2 - 3 | 247 | 99 | |
XM_008773356.1 | XP_008771578.1 | 2 - 3 | 247 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |