Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ell2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760603.1 | XP_008758825.1 | 5 - 6 | 1479 | 159 | |
XM_008760604.1 | XP_008758826.1 | 5 - 6 | 765 | 159 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |