Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xrcc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006999.1 | NP_001007000.1 | 5 - 6 | 657 | 68 | |
XM_006231751.1 | XP_006231813.1 | 5 - 6 | 826 | 68 | ||
XM_008760636.1 | XP_008758858.1 | 5 - 6 | 788 | 68 | ||
XM_008760637.1 | XP_008758859.1 | 5 - 6 | 853 | 68 | ||
XM_008760638.1 | XP_008758860.1 | 5 - 6 | 788 | 68 | ||
GenBank mRNA | BC083848.1 | - | 5 - 6 | 657 | 7122361 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |