Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ercc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106228.1 | NP_001099698.1 | 2 - 3 | 220 | 75 | |
XM_006228395.2 | XP_006228457.1 | 2 - 3 | 238 | 75 | ||
XM_006228396.1 | XP_006228458.1 | 2 - 3 | 198 | 75 | ||
GenBank mRNA | FQ225470.1 | - | 2 - 3 | 209 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |