Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023976.2 | NP_076466.2 | 4 - 5 | 304 | 124 | |
XM_006228586.2 | XP_006228648.1 | 4 - 5 | 397 | 124 | ||
XM_008759196.1 | XP_008757418.1 | 3 - 4 | 498 | 124 | ||
GenBank mRNA | Z29649.2 | - | 4 - 5 | 304 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |