Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130750.1 | NP_570106.1 | 6 - 7 | 705 | 124 | |
GenBank mRNA | AF461260.1 | - | 6 - 7 | 705 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |