Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec11c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153628.2 | NP_705892.1 | 1 - 2 | 139 | 78 | |
GenBank mRNA | AB022714.2 | - | 1 - 2 | 140 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Rattus norvegicus Probe spans exons |