Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019222.1 | NP_062095.1 | 3 - 4 | 343 | 113 | |
XM_006230773.2 | XP_006230835.1 | 3 - 4 | 462 | 113 | ||
XM_006230774.2 | XP_006230836.1 | 3 - 4 | 427 | 113 | ||
GenBank mRNA | AJ006064.1 | - | 3 - 4 | 343 | 113 | |
BC061558.1 | - | 3 - 4 | 396 | 6919414 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |