Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241646.2 | XP_006241708.1 | 3 - 4 | 540 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons |