Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dscc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_216934.8 | XP_216934.3 | 8 - 9 | 1347 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus Probe spans exons |