Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013172.1 | NP_037304.1 | 2 - 3 | 705 | 64 | |
GenBank mRNA | M27151.1 | - | 2 - 3 | 655 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |