Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004283.1 | NP_001004283.1 | 4 - 5 | 397 | 57 | |
XM_006241938.2 | XP_006242000.1 | 4 - 5 | 511 | 57 | ||
GenBank mRNA | BC079005.1 | - | 4 - 5 | 397 | 7113045 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |