Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inhbe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031815.2 | NP_114003.2 | 1 - 2 | 295 | 65 | |
GenBank mRNA | AF089825.1 | - | 486 | 65 | ||
AF140032.1 | - | 1 - 2 | 295 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |