Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133398.1 | NP_596889.1 | 2 - 3 | 790 | 81 | |
XM_006241520.2 | XP_006241582.1 | 2 - 3 | 2209 | 81 | ||
XM_006241521.2 | XP_006241583.1 | 2 - 3 | 2210 | 81 | ||
XM_006241523.2 | XP_006241585.1 | 2 - 3 | 2207 | 81 | ||
XM_006241524.2 | XP_006241586.1 | 2 - 3 | 2207 | 81 | ||
XM_006241525.2 | XP_006241587.1 | 2 - 3 | 2208 | 81 | ||
GenBank mRNA | AF100421.2 | - | 2 - 3 | 790 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Rattus norvegicus Probe spans exons |