Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012730.1 | NP_036862.1 | 3 - 4 | 638 | 71 | |
GenBank mRNA | AB008423.1 | - | 3 - 4 | 514 | 71 | |
BC078897.1 | - | 3 - 4 | 573 | 7109496 | 71 | |
FQ218496.1 | - | 3 - 4 | 579 | 71 | ||
M16655.1 | - | 3 - 4 | 564 | 71 | ||
M22330.1 | - | 3 - 4 | 638 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |