Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyth4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130577.1 | NP_001124049.1 | 9 - 10 | 907 | 91 | |
XM_006242031.2 | XP_006242093.1 | 10 - 11 | 994 | 91 | ||
XM_006242033.2 | XP_006242095.1 | 9 - 10 | 911 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |