Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc90b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024885.1 | NP_001020056.1 | 1 - 2 | 398 | 77 | |
GenBank mRNA | BC097480.1 | - | 1 - 2 | 398 | 7458611 | 77 |
FQ215669.1 | - | 1 - 2 | 100 | 77 | ||
FQ216256.1 | - | 1 - 2 | 102 | 77 | ||
FQ218235.1 | - | 1 - 2 | 113 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |