Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022590.2 | NP_072112.2 | 10 - 11 | 1281 | 54 | |
GenBank mRNA | AB070849.1 | - | 9 - 10 | 1416 | 54 | |
AB070850.1 | - | 9 - 10 | 1416 | 54 | ||
AB070976.1 | - | 10 - 11 | 1140 | 54 | ||
U29881.1 | - | 10 - 11 | 1281 | 54 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |