Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Insc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106285.1 | NP_001099755.1 | 5 - 6 | 877 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |