Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Folr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106283.1 | NP_001099753.1 | 3 - 4 | 421 | 61 | |
XM_006229858.2 | XP_006229920.1 | 2 - 3 | 301 | 61 | ||
XM_006229859.2 | XP_006229921.1 | 3 - 4 | 503 | 61 | ||
XM_008759703.1 | XP_008757925.1 | 2 - 3 | 336 | 61 | ||
GenBank mRNA | FQ211323.1 | - | 2 - 3 | 268 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Rattus norvegicus Probe spans exons |