Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006230186.2 | XP_006230248.1 | 3 - 4 | 1007 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Rattus norvegicus Probe spans exons |