Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130411.2 | NP_569095.1 | 7 - 8 | 963 | 75 | |
XM_006230204.2 | XP_006230266.1 | 7 - 8 | 1047 | 75 | ||
GenBank mRNA | AF416730.1 | - | 6 - 7 | 859 | 75 | |
AF495469.1 | - | 7 - 8 | 963 | 75 | ||
BC086971.1 | - | 7 - 8 | 939 | 7124429 | 75 | |
FQ225360.1 | - | 4 - 5 | 485 | 75 | ||
FQ225522.1 | - | 3 - 4 | 401 | 76 | ||
FQ226290.1 | - | 5 - 6 | 432 | 75 | ||
FQ226379.1 | - | 7 - 8 | 954 | 75 | ||
FQ226941.1 | - | 7 - 8 | 983 | 75 | ||
FQ233306.1 | - | 7 - 8 | 957 | 75 | ||
FQ233390.1 | - | 7 - 8 | 959 | 75 | ||
FQ234750.1 | - | 7 - 8 | 965 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |