Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnq5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134643.2 | NP_001128115.2 | 6 - 7 | 1203 | 84 | |
GenBank mRNA | BF523361.1 | - | 1 - 2 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Ion Channel Rattus norvegicus Probe spans exons |