Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108197.1 | NP_001101667.1 | 4 - 5 | 382 | 75 | |
XM_008766860.1 | XP_008765082.1 | 4 - 5 | 383 | 75 | ||
XM_008766861.1 | XP_008765083.1 | 4 - 5 | 383 | 75 | ||
XM_008766862.1 | XP_008765084.1 | 4 - 5 | 383 | 75 | ||
GenBank mRNA | FQ211123.1 | - | 4 - 5 | 379 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |