Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdca7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025693.1 | NP_001020864.1 | 8 - 9 | 1192 | 63 | |
GenBank mRNA | BC098690.1 | - | 8 - 9 | 1192 | 7386103 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |