Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arfgap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033707.1 | NP_001028879.1 | 5 - 6 | 530 | 86 | |
XM_006234529.2 | XP_006234591.1 | 5 - 6 | 635 | 86 | ||
XM_006234530.2 | XP_006234592.1 | 5 - 6 | 634 | 86 | ||
XM_006234531.2 | XP_006234593.1 | 5 - 6 | 634 | 86 | ||
GenBank mRNA | BC101917.1 | - | 5 - 6 | 530 | 7935829 | 86 |
FQ218185.1 | - | 5 - 6 | 602 | 86 | ||
FQ228393.1 | - | 5 - 6 | 597 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |